Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037519.1 | NP_001032608.1 | 2 | 281 | 69 | |
GenBank mRNA | AY621358.1 | - | 2 | 281 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Both primers and probe map within a single exon |