Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkx1-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_234082.7 | XP_234082.6 | 2 | 1108 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon |