Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001000542.1 | NP_001000542.1 | 1 | 898 | 118 | |
XM_006229890.2 | XP_006229952.1 | 2 | 1215 | 118 | ||
GenBank mRNA | AY834218.1 | - | 1 | 898 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |