Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ier3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212505.2 | NP_997670.1 | 2 | 999 | 156 | |
GenBank mRNA | BC099831.1 | - | 2 | 975 | 7933804 | 156 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |