Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108807.1 | NP_001102277.1 | 3 | 461 | 139 | |
XM_006245414.2 | XP_006245476.1 | 2 | 912 | 139 | ||
XM_006245415.2 | XP_006245477.1 | 2 | 549 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |