Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017497.1 | NP_001017497.1 | 3 | 547 | 141 | |
GenBank mRNA | X77816.1 | - | 3 | 547 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon |