Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U36896.1 | - | 3 | 1328 | 194 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Amplicon spans exons and probe does not span exons |