Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hyal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207616.1 | NP_997499.1 | 4 | 1295 | 99 | |
GenBank mRNA | AB100600.1 | - | 4 | 1295 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |