Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005899.2 | NP_001005899.2 | 2 - 3 | 213 | 146 | |
GenBank mRNA | AY363176.1 | - | 2 - 3 | 259 | 146 | |
AY494060.1 | - | 2 - 3 | 212 | 146 | ||
AY581051.1 | - | 2 - 3 | 213 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |