Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008776.2 | NP_001008776.1 | 2 - 3 | 919 | 123 | |
XM_006240481.2 | XP_006240543.1 | 3 - 4 | 1024 | 123 | ||
GenBank mRNA | AY325135.1 | - | 2 - 3 | 959 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |