Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008899.1 | NP_001008899.1 | 7 - 8 | 1528 | 114 | |
XM_006242436.2 | XP_006242498.1 | 7 - 8 | 2029 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |