Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009419.1 | NP_001009419.1 | 3 - 4 | 558 | 147 | |
GenBank mRNA | BC087599.1 | - | 3 - 4 | 558 | 7313414 | 147 |
FQ230721.1 | - | 2 - 3 | 173 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |