Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikbip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241250.2 | XP_006241312.1 | 2 - 3 | 457 | 108 | |
GenBank mRNA | BC088210.1 | - | 2 - 3 | 361 | 7382451 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |