Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009536.1 | NP_001009536.1 | 4 - 5 | 1152 | 89 | |
GenBank mRNA | BC061749.1 | - | 4 - 5 | 1152 | 5599464 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |