Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fastkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009673.1 | NP_001009673.1 | 6 - 7 | 1450 | 128 | |
XM_006245050.1 | XP_006245112.1 | 7 - 8 | 1607 | 128 | ||
GenBank mRNA | BC088416.1 | - | 6 - 7 | 1450 | 7112913 | 128 |
FQ218822.1 | - | 2 - 3 | 255 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |