Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oasl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009682.1 | NP_001009682.1 | 4 - 5 | 966 | 138 | |
XM_008769327.1 | XP_008767549.1 | 4 - 5 | 1140 | 138 | ||
GenBank mRNA | AY237116.1 | - | 4 - 5 | 966 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |