Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009490.1 | NP_001009490.1 | 1 - 2 | 212 | 134 | |
GenBank mRNA | AY196700.1 | - | 1 - 2 | 212 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |