Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009492.1 | NP_001009492.1 | 2 - 3 | 494 | 153 | |
XM_008769248.1 | XP_008767470.1 | 2 - 3 | 495 | 153 | ||
GenBank mRNA | AY196698.1 | - | 2 - 3 | 494 | 153 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Inventoried Probe spans exons |