Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psbpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207593.2 | NP_997476.2 | 1 - 2 | 85 | 126 | |
GenBank mRNA | BC099748.1 | - | 1 - 2 | 79 | 126 | |
V01256.1 | - | 1 - 2 | 85 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |