Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008760.1 | NP_001008760.1 | 6 - 7 | 1012 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |