Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008808.1 | NP_001008808.1 | 4 - 5 | 808 | 156 | |
XM_008765703.1 | XP_008763925.1 | 4 - 5 | 848 | 156 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |