Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klri2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012648.1 | NP_001012666.1 | 2 - 3 | 331 | 98 | |
GenBank mRNA | AY324871.1 | - | 2 - 3 | 331 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |