Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012748.1 | NP_001012766.1 | 8 - 9 | 1528 | 95 | |
GenBank mRNA | AB121033.1 | - | 8 - 9 | 1528 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |