Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014023.1 | NP_001014045.1 | 6 - 7 | 977 | 74 | |
XM_006229912.2 | XP_006229974.1 | 6 - 7 | 1463 | 74 | ||
GenBank mRNA | BC078926.1 | - | 6 - 7 | 977 | 7106563 | 74 |
FQ225508.1 | - | 1 - 2 | 96 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |