Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014077.1 | NP_001014099.1 | 1 | 2576 | 70 | |
GenBank mRNA | BC081897.1 | - | 1 | 2576 | 7110446 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Both primers and probe map within a single exon |