Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spert | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017506.1 | NP_001017506.1 | 1 - 2 | 124 | 124 | |
GenBank mRNA | BC079300.1 | - | 1 - 2 | 124 | 7134526 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |