Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hyal5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024321.1 | NP_001019492.1 | 2 - 3 | 1263 | 80 | |
XM_006236166.2 | XP_006236228.1 | 3 - 4 | 1381 | 80 | ||
XM_008762806.1 | XP_008761028.1 | 4 - 5 | 1609 | 80 | ||
XM_008762807.1 | XP_008761029.1 | 3 - 4 | 1516 | 80 | ||
XM_008762808.1 | XP_008761030.1 | 3 - 4 | 1431 | 80 | ||
GenBank mRNA | BC079447.1 | - | 4 - 5 | 1586 | 80 | |
BC091219.1 | - | 2 - 3 | 1263 | 7112082 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |