Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024332.1 | NP_001019503.1 | 5 - 6 | 622 | 131 | |
GenBank mRNA | BC081947.1 | - | 5 - 6 | 622 | 7121584 | 131 |
FQ220718.1 | - | 5 - 6 | 614 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |