Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp11b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012538.2 | NP_036670.2 | 4 - 5 | 807 | 103 | |
XM_008765540.1 | XP_008763762.1 | 4 - 5 | 845 | 103 | ||
XM_008765541.1 | XP_008763763.1 | 4 - 5 | 845 | 103 | ||
XM_008765542.1 | XP_008763764.1 | 4 - 5 | 845 | 103 | ||
GenBank mRNA | D00568.1 | - | 4 - 5 | 805 | 103 | |
X52766.1 | - | 4 - 5 | 801 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |