Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008823.1 | NP_001008823.1 | 6 | 1058 | 116 | |
XM_006247390.2 | XP_006247452.1 | 6 | 1646 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Both primers and probe map within a single exon |