Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tax1bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025419.1 | NP_001020590.1 | 1 - 2 | 73 | 57 | |
XM_006246761.2 | XP_006246823.1 | 1 - 2 | 280 | 57 | ||
GenBank mRNA | BC097976.1 | - | 1 - 2 | 73 | 7442643 | 57 |
FQ215653.1 | - | 1 - 2 | 92 | 57 | ||
FQ219871.1 | - | 1 - 2 | 93 | 57 | ||
FQ220071.1 | - | 1 - 2 | 92 | 57 | ||
FQ220113.1 | - | 1 - 2 | 92 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons |