Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025654.1 | NP_001020825.1 | 1 - 2 | 65 | 64 | |
GenBank mRNA | DQ058654.1 | - | 1 - 2 | 65 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons |