Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025658.1 | NP_001020829.1 | 3 - 4 | 767 | 106 | |
XM_006240226.2 | XP_006240288.1 | 5 - 6 | 970 | 106 | ||
XM_006240227.2 | XP_006240289.1 | 5 - 6 | 1014 | 106 | ||
XM_008764744.1 | XP_008762966.1 | 4 - 5 | 885 | 106 | ||
XM_008764745.1 | XP_008762967.1 | 3 - 4 | 490 | 106 | ||
GenBank mRNA | BC097422.1 | - | 3 - 4 | 767 | 7454996 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |