Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defa8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033077.1 | NP_001028249.1 | 1 - 2 | 170 | 76 | |
GenBank mRNA | AY623752.1 | - | 1 - 2 | 170 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |