Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RT1-M6-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008852.1 | NP_001008852.1 | 1 - 2 | 88 | 140 | |
XM_008772698.1 | XP_008770920.1 | 1 - 2 | 89 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |