Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047095.1 | NP_001040560.1 | 2 - 3 | 284 | 58 | |
GenBank mRNA | AB256043.1 | - | 2 - 3 | 284 | 58 | |
AM258958.1 | - | 2 - 3 | 284 | 58 | ||
BC158562.1 | - | 2 - 3 | 288 | 9088902 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |