Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pacrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077677.1 | NP_001071145.1 | 4 - 5 | 814 | 103 | |
GenBank mRNA | BC127534.1 | - | 4 - 5 | 814 | 7133203 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |