Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmox2-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045198.1 | 1 | 2098 | 103 | ||
GenBank mRNA | AF058787.1 | - | 2059 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |