Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024823.4 | NP_001019994.4 | 1 | 1806 | 145 | |
NM_021845.5 | NP_068617.6 | 1 | 1806 | 145 | ||
GenBank mRNA | X84047.1 | - | 1 | 954 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |