Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mageb18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044246.1 | NP_001037711.1 | 2 | 1057 | 144 | |
GenBank mRNA | BC097441.1 | - | 2 | 1057 | 7456024 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon |