Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047869.1 | NP_001041334.1 | 1 | 3570 | 106 | |
XM_008764656.1 | XP_008762878.1 | 1 | 3590 | 106 | ||
GenBank mRNA | AY387084.1 | - | 1 | 3570 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |