Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcdc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114621.1 | NP_001108093.1 | 9 - 10 | 1080 | 86 | |
XM_008762082.1 | XP_008760304.1 | 10 - 11 | 2279 | 86 | ||
XM_008762083.1 | XP_008760305.1 | 4 - 5 | 1011 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |