Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC287004 | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110636.1 | 1 - 2 | 238 | 102 | |||
NR_110638.1 | 1 - 2 | 238 | 102 | ||||
NR_110640.1 | 1 - 2 | 238 | 102 | ||||
GenBank mRNA | AY035343.1 | - | LIT1742 PINC |
1 - 2 | 235 | 102 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | protein unc-80 homolog | LOC103693241 | -54.4kb | Sense | Chr. 9 - 72663790 - 72665685 on NCBI Build Rnor_6.0 | |
Downstream | tensin 1 | Tns1 | Tns | 494.4kb | Sense | Chr. 9 - 81191792 - 81401027 on NCBI Build Rnor_6.0 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |