Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271731.1 | NP_001258660.1 | 4 - 5 | 501 | 123 | |
GenBank mRNA | CB691171.1 | - | 2 - 3 | 99 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Inventoried Probe spans exons |