Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135090.1 | NP_001128562.1 | 1 | 2042 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Both primers and probe map within a single exon |