Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agmo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135899.1 | NP_001129371.1 | 7 - 8 | 879 | 132 | |
GenBank mRNA | BC127545.1 | - | 7 - 8 | 879 | 7321962 | 132 |
FQ211046.1 | - | 7 - 8 | 935 | 132 | ||
FQ219104.1 | - | 7 - 8 | 923 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |