Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctdnep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100494.1 | NP_001093964.1 | 1 - 2 | 308 | 83 | |
XM_008767804.1 | XP_008766026.1 | 1 - 2 | 337 | 83 | ||
GenBank mRNA | BU671232.1 | - | 1 - 2 | 308 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |