Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168630.1 | NP_001162101.1 | 6 - 7 | 1251 | 99 | |
XM_006232076.2 | XP_006232138.1 | 8 - 9 | 1475 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |