Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169149.1 | NP_001162620.1 | 5 - 6 | 2352 | 75 | |
XM_006228188.1 | XP_006228250.1 | 4 - 5 | 2187 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |