Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007744.1 | NP_001007745.1 | 3 - 4 | 643 | 70 | |
XM_006233981.2 | XP_006234043.1 | 3 - 4 | 1278 | 70 | ||
GenBank mRNA | BC078943.1 | - | 3 - 4 | 643 | 7109352 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |